Learn More
Meso Scale Discovery R-PLEX Human VILIP-1 ASY
SDP
Catalog No. 5000080286
Encompass
The R-PLEX Human VILIP-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures VILIP-1 levels in human serum plasma (EDTA) and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product enables the quantification of VILIP-1 for research use only
Catalog No. 50-000-80286 Supplier Meso Scale Discovery Supplier No. K1512FR2
The R-PLEX Human VILIP-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures VILIP-1 levels in human serum plasma (EDTA) and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product enables the quantification of VILIP-1 for research use only