Learn More
Meso Scale Discovery R-PLEX Human VILIP-1 ASYQP
SDP
Catalog No. 5000080210
Encompass
The R-PLEX Human VILIP-1 Assay Kit from Meso Scale Discovery measures VILIP-1 in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture and detection antibodies buffers blocker solutions calibrator and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It requires 50 L per well and can assay 46 samples in duplicate with a total time of 3 5 hours
Catalog No. 50-000-80210 Supplier Meso Scale Discovery Supplier No. K1512FR22
Add to Cart
Add to Cart
The R-PLEX Human VILIP-1 Assay Kit from Meso Scale Discovery measures VILIP-1 in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture and detection antibodies buffers blocker solutions calibrator and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It requires 50 L per well and can assay 46 samples in duplicate with a total time of 3 5 hours