Learn More
Meso Scale Discovery R-PLEX Human Visfatin ASY
SDP
Catalog No. 5000080582
Encompass
The Meso Scale Discovery R-PLEX Human Visfatin Assay measures visfatin in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity a 5-log dynamic range and low sample volume needs Fully quantitative with an 8-point calibration curve it is lyophilized for stability The R-PLEX platform allows multiplexing with other biomarkers This assay provides robust data for research in obesity diabetes metabolic disease and immunology
Catalog No. 50-000-80582 Supplier Meso Scale Discovery Supplier No. K151Q5R2
The Meso Scale Discovery R-PLEX Human Visfatin Assay measures visfatin in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity a 5-log dynamic range and low sample volume needs Fully quantitative with an 8-point calibration curve it is lyophilized for stability The R-PLEX platform allows multiplexing with other biomarkers This assay provides robust data for research in obesity diabetes metabolic disease and immunology