Learn More
Meso Scale Discovery R-PLEX Human VISTA/B7-H5 ASY
SDP
Catalog No. 5000080426
Encompass
The R-PLEX Human Vista B7-H5 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-5 IL-6 IL-8 TNF- IFN- IL-10 IL-17A IL-17E/IL-25 IL-17F IL-21 IL-22 IL-23 IL-27 IL-31 and GM-CSF It uses electrochemiluminescent detection offers high sensitivity an 8-log dynamic range and requires low sample volumes The kit includes a 96-well plate detection antibodies buffers diluent calibrators and controls Compatible with the MESO QuickPlex SQ 120 it provides results in 3 5 hours
Catalog No. 50-000-80426 Supplier Meso Scale Discovery Supplier No. K151V3R2
The R-PLEX Human Vista B7-H5 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-5 IL-6 IL-8 TNF- IFN- IL-10 IL-17A IL-17E/IL-25 IL-17F IL-21 IL-22 IL-23 IL-27 IL-31 and GM-CSF It uses electrochemiluminescent detection offers high sensitivity an 8-log dynamic range and requires low sample volumes The kit includes a 96-well plate detection antibodies buffers diluent calibrators and controls Compatible with the MESO QuickPlex SQ 120 it provides results in 3 5 hours