Learn More
Meso Scale Discovery R-PLEX Human VTCN1/B7-H4 ASYQP
SDP
Catalog No. 5000080191
Encompass
The R-PLEX Human VTCN1 (B7-H4) Assay is a multiplex immunoassay panel that measures VTCN1 (B7-H4) in human serum plasma (EDTA heparin) and cell culture supernatant samples This assay uses electrochemiluminescence (ECL) detection technology on the Meso Scale Discovery (MSD) platform It allows for the quantitative measurement of VTCN1 with high sensitivity and wide dynamic range This assay provides a reproducible and robust tool for studying VTCN1 biology and evaluating VTCN1 as a biomarker across disease states including cancer It requires 50 L sample volume and can process up to 80 samples in duplicates per plate
Catalog No. 50-000-80191 Supplier Meso Scale Discovery Supplier No. K151ALMR22
The R-PLEX Human VTCN1 (B7-H4) Assay is a multiplex immunoassay panel that measures VTCN1 (B7-H4) in human serum plasma (EDTA heparin) and cell culture supernatant samples This assay uses electrochemiluminescence (ECL) detection technology on the Meso Scale Discovery (MSD) platform It allows for the quantitative measurement of VTCN1 with high sensitivity and wide dynamic range This assay provides a reproducible and robust tool for studying VTCN1 biology and evaluating VTCN1 as a biomarker across disease states including cancer It requires 50 L sample volume and can process up to 80 samples in duplicates per plate