Learn More
Meso Scale Discovery R-PLEX Human ZnT8 ASY
SDP
Catalog No. 5000080150
Encompass
The Meso Scale Discovery R-Plex Human ZNT8 Assay is a multiplex immunoassay that measures zinc transporter 8 (ZNT8) autoantibodies in human serum plasma and dried blood spot samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides a quantitative measurement of ZNT8 autoantibody levels ZNT8 autoantibodies are biomarkers that indicate increased risk for type 1 diabetes (T1D) This assay kit includes a 96-well plate coated with ZNT8 antigen detection antibody read buffer sample diluent calibrators and controls The total assay time is approximately 3 5 hours This product can aid in understanding ZNT8 autoimmunity and T1D risk assessment
Catalog No. 50-000-80150 Supplier Meso Scale Discovery Supplier No. K151AKDR2
The Meso Scale Discovery R-Plex Human ZNT8 Assay is a multiplex immunoassay that measures zinc transporter 8 (ZNT8) autoantibodies in human serum plasma and dried blood spot samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides a quantitative measurement of ZNT8 autoantibody levels ZNT8 autoantibodies are biomarkers that indicate increased risk for type 1 diabetes (T1D) This assay kit includes a 96-well plate coated with ZNT8 antigen detection antibody read buffer sample diluent calibrators and controls The total assay time is approximately 3 5 hours This product can aid in understanding ZNT8 autoimmunity and T1D risk assessment