Learn More
Meso Scale Discovery R-PLEX Human ZnT8 ASYQP
SDP
Catalog No. 5000081787
Encompass
The Meso Scale Discovery R-Plex Human ZNT8 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Zinc Transporter 8 (ZNT8) autoantibodies in human serum plasma and other biological matrices This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with ZNT8 antigen detection antibody read buffer calibrator and controls It can be used for research purposes to study type 1 diabetes and other autoimmune conditions involving ZNT8 autoantibodies The kit has high lot-to-lot consistency
Catalog No. 50-000-81787 Supplier Meso Scale Discovery Supplier No. K151AKDR22
The Meso Scale Discovery R-Plex Human ZNT8 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Zinc Transporter 8 (ZNT8) autoantibodies in human serum plasma and other biological matrices This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with ZNT8 antigen detection antibody read buffer calibrator and controls It can be used for research purposes to study type 1 diabetes and other autoimmune conditions involving ZNT8 autoantibodies The kit has high lot-to-lot consistency