Learn More
Meso Scale Discovery R-PLEX Mouse Adiponectin ASYQP
SDP
Catalog No. 5000082426
Encompass
The Meso Scale Discovery R-PLEX Mouse Adiponectin Assay is a multiplex immunoassay panel that allows simultaneous measurement of adiponectin in mouse samples such as serum plasma and tissue culture media This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 QuickPlex SQ 120i MESO QuickPlex SQ 120 and V-PLEX Plus The assay requires 50 L or less of sample and can quantitate adiponectin over a range of 156 pg/mL to 100 000 pg/mL
Catalog No. 50-000-82426 Supplier Meso Scale Discovery Supplier No. K153YQR22
The Meso Scale Discovery R-PLEX Mouse Adiponectin Assay is a multiplex immunoassay panel that allows simultaneous measurement of adiponectin in mouse samples such as serum plasma and tissue culture media This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 QuickPlex SQ 120i MESO QuickPlex SQ 120 and V-PLEX Plus The assay requires 50 L or less of sample and can quantitate adiponectin over a range of 156 pg/mL to 100 000 pg/mL