Learn More
Meso Scale Discovery R-PLEX Mouse C1q receptor ASY
SDP
Catalog No. 5000079101
Encompass
The Meso Scale Discovery Mouse C1q Receptor R-PLEX Assay is a multiplex immunoassay that quantitatively measures C1q Receptor in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120 The assay has a detection range of 50-100 000 pg/mL and the total assay time is approximately 3 5 hours This assay allows for the rapid reproducible quantification of C1q Receptor from mouse samples
Catalog No. 50-000-79101 Supplier Meso Scale Discovery Supplier No. K152S1R2
The Meso Scale Discovery Mouse C1q Receptor R-PLEX Assay is a multiplex immunoassay that quantitatively measures C1q Receptor in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120 The assay has a detection range of 50-100 000 pg/mL and the total assay time is approximately 3 5 hours This assay allows for the rapid reproducible quantification of C1q Receptor from mouse samples