Learn More
Meso Scale Discovery R-PLEX Mouse CD14 ASY
SDP
Catalog No. 5000079359
Encompass
The R-PLEX Mouse CD14 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD14 in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This 5-plex assay kit includes a CD14-specific antibody coated plate and calibrator diluent that allow for the accurate quantification of CD14 The assay takes approximately 4 hours to complete and is suitable for measuring CD14 levels during immunology and inflammation studies in mice
Catalog No. 50-000-79359 Supplier Meso Scale Discovery Supplier No. K152T1R2
The R-PLEX Mouse CD14 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD14 in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This 5-plex assay kit includes a CD14-specific antibody coated plate and calibrator diluent that allow for the accurate quantification of CD14 The assay takes approximately 4 hours to complete and is suitable for measuring CD14 levels during immunology and inflammation studies in mice