Learn More
Meso Scale Discovery R-PLEX Mouse Clusterin ASY
SDP
Catalog No. 5000081452
Encompass
The Meso Scale Discovery R-PLEX Mouse Clusterin Assay is a multiplex immunoassay panel that allows simultaneous measurement of Clusterin in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay is provided in a convenient all-in-one kit including calibrators controls assay diluent read buffer and plate The R-PLEX Mouse Clusterin Assay allows for efficient and accurate quantification of the Clusterin biomarker
Catalog No. 50-000-81452 Supplier Meso Scale Discovery Supplier No. K152E0R2
The Meso Scale Discovery R-PLEX Mouse Clusterin Assay is a multiplex immunoassay panel that allows simultaneous measurement of Clusterin in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay is provided in a convenient all-in-one kit including calibrators controls assay diluent read buffer and plate The R-PLEX Mouse Clusterin Assay allows for efficient and accurate quantification of the Clusterin biomarker