Learn More
Meso Scale Discovery R-PLEX Mouse Clusterin ASYQP
SDP
Catalog No. 5000082106
Encompass
The Meso Scale Discovery R-PLEX Mouse Clusterin Assay is a multiplex immunoassay for measuring Clusterin in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescence technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes calibrators controls diluent read buffer and 96-well plates with capture antibodies for quick reproducible results It is compatible with MSD s SECTOR Imager plate reader providing a sensitive method for Clusterin measurement
Catalog No. 50-000-82106 Supplier Meso Scale Discovery Supplier No. K152E0R22
The Meso Scale Discovery R-PLEX Mouse Clusterin Assay is a multiplex immunoassay for measuring Clusterin in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescence technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes calibrators controls diluent read buffer and 96-well plates with capture antibodies for quick reproducible results It is compatible with MSD s SECTOR Imager plate reader providing a sensitive method for Clusterin measurement