Learn More
Meso Scale Discovery R-PLEX Mouse CRP ASYQP
SDP
Catalog No. 5000082351
Encompass
The R-PLEX Mouse CRP Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in mouse serum plasma and tissue culture media The assay uses MSD s proprietary SECTOR Imager and provides a sensitive accurate and reproducible method to detect CRP levels between 0 8-500 ng/mL The assay can be used to monitor inflammation and infection in mouse models It utilizes a sandwich immunoassay format with an MSD SULFO-TAG label that emits light upon electrochemical stimulation The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent
Catalog No. 50-000-82351 Supplier Meso Scale Discovery Supplier No. K1525KR22
The R-PLEX Mouse CRP Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in mouse serum plasma and tissue culture media The assay uses MSD s proprietary SECTOR Imager and provides a sensitive accurate and reproducible method to detect CRP levels between 0 8-500 ng/mL The assay can be used to monitor inflammation and infection in mouse models It utilizes a sandwich immunoassay format with an MSD SULFO-TAG label that emits light upon electrochemical stimulation The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent