Learn More
Meso Scale Discovery R-PLEX Mouse EGF ASY
SDP
Catalog No. 5000081451
Encompass
The R-Plex Mouse EGF Assay Sector 5-PL K152ALHR is a multiplex immunoassay kit for measuring mouse epidermal growth factor (EGF) in biological samples It uses electrochemiluminescence (ECL) detection technology and provides reagents for 96 tests in a 96-well plate format The assay has a typical sensitivity of 0 3 pg/mL and requires 10 L of sample The kit includes a plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators controls assay and wash buffers signal enhancer solutions and a plate seal This quantitative assay allows high-throughput profiling of mouse EGF with accuracy and reproducibility
Catalog No. 50-000-81451 Supplier Meso Scale Discovery Supplier No. K152ALHR2
The R-Plex Mouse EGF Assay Sector 5-PL K152ALHR is a multiplex immunoassay kit for measuring mouse epidermal growth factor (EGF) in biological samples It uses electrochemiluminescence (ECL) detection technology and provides reagents for 96 tests in a 96-well plate format The assay has a typical sensitivity of 0 3 pg/mL and requires 10 L of sample The kit includes a plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators controls assay and wash buffers signal enhancer solutions and a plate seal This quantitative assay allows high-throughput profiling of mouse EGF with accuracy and reproducibility