Learn More
Meso Scale Discovery R-PLEX Mouse EGF ASYQP
SDP
Catalog No. 5000078484
Encompass
The R-Plex Mouse EGF Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring mouse epidermal growth factor (EGF) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The assay is specific with minimal cross-reactivity and includes a 96-well plate antibodies buffers diluent calibrators and controls It tests up to 80 samples in duplicates within 3 5 hours
Catalog No. 50-000-78484 Supplier Meso Scale Discovery Supplier No. K152ALHR22
The R-Plex Mouse EGF Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring mouse epidermal growth factor (EGF) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The assay is specific with minimal cross-reactivity and includes a 96-well plate antibodies buffers diluent calibrators and controls It tests up to 80 samples in duplicates within 3 5 hours