Learn More
Meso Scale Discovery R-PLEX Mouse Fractalkine ASY
SDP
Catalog No. 5000080448
Encompass
The R-PLEX Mouse Fractalkine Assay from Meso Scale Discovery is a multiplex immunoassay that measures Fractalkine levels in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity and wide dynamic range A proprietary diluent is provided for optimal sample dilution Each kit provides sufficient reagents to assay one 96-well plate This quantitative assay provides accurate and reproducible results with minimal sample volume requirements It offers a streamlined workflow and rapid results
Catalog No. 50-000-80448 Supplier Meso Scale Discovery Supplier No. K152V1R2
The R-PLEX Mouse Fractalkine Assay from Meso Scale Discovery is a multiplex immunoassay that measures Fractalkine levels in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity and wide dynamic range A proprietary diluent is provided for optimal sample dilution Each kit provides sufficient reagents to assay one 96-well plate This quantitative assay provides accurate and reproducible results with minimal sample volume requirements It offers a streamlined workflow and rapid results