Learn More
Meso Scale Discovery R-PLEX Mouse Fractalkine ASYQP
SDP
Catalog No. 5000078985
Encompass
The R-PLEX Mouse Fractalkine Assay from Meso Scale Discovery is a multiplex immunoassay that allows for the simultaneous quantitative measurement of fractalkine in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay can be run on MSD s QuickPlex SQ 120 instrument and data analysis is performed using MSD s Discovery Workbench software This product enables the reliable quantification of fractalkine which plays important roles in immune cell trafficking and inflammatory processes
Catalog No. 50-000-78985 Supplier Meso Scale Discovery Supplier No. K152V1R22
The R-PLEX Mouse Fractalkine Assay from Meso Scale Discovery is a multiplex immunoassay that allows for the simultaneous quantitative measurement of fractalkine in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay can be run on MSD s QuickPlex SQ 120 instrument and data analysis is performed using MSD s Discovery Workbench software This product enables the reliable quantification of fractalkine which plays important roles in immune cell trafficking and inflammatory processes