Learn More
Meso Scale Discovery R-PLEX Mouse G-CSF ASY
SDP
Catalog No. 5000080015
Encompass
The R-PLEX Mouse G-CSF Assay is a multiplex immunoassay panel that measures G-CSF in mouse samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and is designed for use on Meso Scale Discovery (MSD) instrumentation platforms The assay allows sensitive accurate and rapid quantification of G-CSF levels It requires a small sample volume and provides a dynamic range of up to 3 logs The panel is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies calibrators and controls The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80015 Supplier Meso Scale Discovery Supplier No. K152AJLR2
The R-PLEX Mouse G-CSF Assay is a multiplex immunoassay panel that measures G-CSF in mouse samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and is designed for use on Meso Scale Discovery (MSD) instrumentation platforms The assay allows sensitive accurate and rapid quantification of G-CSF levels It requires a small sample volume and provides a dynamic range of up to 3 logs The panel is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies calibrators and controls The assay takes approximately 3 5 hours to complete