Learn More
Meso Scale Discovery R-PLEX Mouse G-CSF ASYQP
SDP
Catalog No. 5000081380
Encompass
The R-PLEX Mouse G-CSF Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of granulocyte colony stimulating factor (G-CSF) in mouse samples such as serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81380 Supplier Meso Scale Discovery Supplier No. K152AJLR22
The R-PLEX Mouse G-CSF Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of granulocyte colony stimulating factor (G-CSF) in mouse samples such as serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL