Learn More
Meso Scale Discovery R-PLEX Mouse HGF ASY
SDP
Catalog No. 5000081491
Encompass
The R-PLEX Mouse HGF Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of hepatocyte growth factor (HGF) in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies specific for mouse HGF detection antibodies labeled with an ECL compound and calibrator diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81491 Supplier Meso Scale Discovery Supplier No. K152ALJR2
The R-PLEX Mouse HGF Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of hepatocyte growth factor (HGF) in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies specific for mouse HGF detection antibodies labeled with an ECL compound and calibrator diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL