Learn More
Meso Scale Discovery R-PLEX Mouse HGF ASYQP
SDP
Catalog No. 5000082358
Encompass
The R-PLEX Mouse HGF Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring hepatocyte growth factor (HGF) in mouse samples like serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies for mouse HGF ECL-labeled detection antibodies and calibrators for a standard curve The assay detects HGF from 1 15 pg/mL to 10 000 pg/mL
Catalog No. 50-000-82358 Supplier Meso Scale Discovery Supplier No. K152ALJR22
The R-PLEX Mouse HGF Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring hepatocyte growth factor (HGF) in mouse samples like serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies for mouse HGF ECL-labeled detection antibodies and calibrators for a standard curve The assay detects HGF from 1 15 pg/mL to 10 000 pg/mL