Learn More
Meso Scale Discovery R-PLEX Mouse ICAM-1 ASY
SDP
Catalog No. 5000080017
Encompass
The R-PLEX Mouse ICAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures mouse Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can assay up to 80 samples in duplicates in 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and positive control This assay allows rapid reproducible quantification of mouse ICAM-1
Catalog No. 50-000-80017 Supplier Meso Scale Discovery Supplier No. K1527NR2
The R-PLEX Mouse ICAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures mouse Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can assay up to 80 samples in duplicates in 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and positive control This assay allows rapid reproducible quantification of mouse ICAM-1