Learn More
Meso Scale Discovery R-PLEX Mouse ICAM-1 ASYQP
SDP
Catalog No. 5000079251
Encompass
The R-PLEX Mouse ICAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures mouse Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can assay up to 80 samples in duplicates in 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and positive control This assay allows rapid reproducible quantification of mouse ICAM-1 for research use
Catalog No. 50-000-79251 Supplier Meso Scale Discovery Supplier No. K1527NR22
The R-PLEX Mouse ICAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures mouse Intercellular Adhesion Molecule 1 (ICAM-1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can assay up to 80 samples in duplicates in 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and positive control This assay allows rapid reproducible quantification of mouse ICAM-1 for research use