Learn More
Meso Scale Discovery R-PLEX Mouse IL-1 ASY
SDP
Catalog No. 5000081347
Encompass
The R-PLEX Mouse IL-1alpha Assay from Meso Scale Discovery measures mouse interleukin 1 alpha (IL-1 ) in various biological samples using electrochemiluminescence (ECL) detection It offers high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The assay quantifies IL-1 concentrations from 0 05 to 10 000 pg/mL with high specificity The kit includes a 96-well plate antibodies calibrators controls diluent buffers and seals enabling rapid reproducible quantification for research
Catalog No. 50-000-81347 Supplier Meso Scale Discovery Supplier No. K1521YR2
Add to Cart
Add to Cart
The R-PLEX Mouse IL-1alpha Assay from Meso Scale Discovery measures mouse interleukin 1 alpha (IL-1 ) in various biological samples using electrochemiluminescence (ECL) detection It offers high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The assay quantifies IL-1 concentrations from 0 05 to 10 000 pg/mL with high specificity The kit includes a 96-well plate antibodies calibrators controls diluent buffers and seals enabling rapid reproducible quantification for research