Learn More
Meso Scale Discovery R-PLEX Mouse IL-7 ASY
SDP
Catalog No. 5000079260
Encompass
The R-PLEX Mouse IL-7 Assay from Meso Scale Discovery is a multiplex immunoassay for measuring mouse interleukin 7 (IL-7) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs It quantifies IL-7 from 0 05 to 10 000 pg/mL with just 25 L of sample The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators diluent wash buffer read buffer and plate seals ensuring rapid and reliable IL-7 quantification
Catalog No. 50-000-79260 Supplier Meso Scale Discovery Supplier No. K152AJAR2
The R-PLEX Mouse IL-7 Assay from Meso Scale Discovery is a multiplex immunoassay for measuring mouse interleukin 7 (IL-7) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs It quantifies IL-7 from 0 05 to 10 000 pg/mL with just 25 L of sample The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators diluent wash buffer read buffer and plate seals ensuring rapid and reliable IL-7 quantification