Learn More
Meso Scale Discovery R-PLEX Mouse IL-7 ASYQP
SDP
Catalog No. 5000082357
Encompass
The R-PLEX Mouse IL-7 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for quantifying mouse interleukin 7 (IL-7) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies for mouse IL-7 ECL-labeled detection antibodies and calibrator diluents for a standard curve The assay takes about 3 5 hours and quantifies IL-7 concentrations from 1 53 to 1000 pg/mL using 25 L samples
Catalog No. 50-000-82357 Supplier Meso Scale Discovery Supplier No. K152AJAR22
The R-PLEX Mouse IL-7 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for quantifying mouse interleukin 7 (IL-7) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies for mouse IL-7 ECL-labeled detection antibodies and calibrator diluents for a standard curve The assay takes about 3 5 hours and quantifies IL-7 concentrations from 1 53 to 1000 pg/mL using 25 L samples