Learn More
Meso Scale Discovery R-PLEX Mouse M-CSF ASY
SDP
Catalog No. 5000078483
Encompass
The R-PLEX Mouse M-CSF Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring mouse macrophage colony stimulating factor (M-CSF) in various biological fluids This ECL-based assay uses MULTI-ARRAY technology and includes a pre-coated 96-well plate calibrators controls diluent detection antibody read buffer and seals It has a sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL The assay offers high sensitivity low sample volume and quick results
Catalog No. 50-000-78483 Supplier Meso Scale Discovery Supplier No. K152AJRR2
The R-PLEX Mouse M-CSF Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring mouse macrophage colony stimulating factor (M-CSF) in various biological fluids This ECL-based assay uses MULTI-ARRAY technology and includes a pre-coated 96-well plate calibrators controls diluent detection antibody read buffer and seals It has a sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL The assay offers high sensitivity low sample volume and quick results