Learn More
Meso Scale Discovery R-PLEX Mouse M-CSF ASYQP
SDP
Catalog No. 5000082385
Encompass
The R-PLEX Mouse M-CSF Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of mouse Macrophage Colony-Stimulating Factor (M-CSF) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less of sample per well and can perform up to 80 tests across two plates It includes a plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls
Catalog No. 50-000-82385 Supplier Meso Scale Discovery Supplier No. K152AJRR22
The R-PLEX Mouse M-CSF Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of mouse Macrophage Colony-Stimulating Factor (M-CSF) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less of sample per well and can perform up to 80 tests across two plates It includes a plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls