Learn More
Meso Scale Discovery R-PLEX Mouse MIG ASYQP
SDP
Catalog No. 5000079728
Encompass
The R-PLEX Mouse MIG Assay from Meso Scale Discovery is a multiplex immunoassay kit for measuring mouse IFN- IL-10 IL-12p70 IL-1 IL-2 IL-4 IL-5 IL-6 KC/GRO and TNF in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5-logs and low sample volume requirements This 10-plex assay efficiently profiles multiple mouse inflammatory cytokines and chemokines from a single small-volume sample
Catalog No. 50-000-79728 Supplier Meso Scale Discovery Supplier No. K1520IR22
The R-PLEX Mouse MIG Assay from Meso Scale Discovery is a multiplex immunoassay kit for measuring mouse IFN- IL-10 IL-12p70 IL-1 IL-2 IL-4 IL-5 IL-6 KC/GRO and TNF in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5-logs and low sample volume requirements This 10-plex assay efficiently profiles multiple mouse inflammatory cytokines and chemokines from a single small-volume sample