Learn More
Meso Scale Discovery R-PLEX Mouse MMP-3 total ASY
SDP
Catalog No. 5000078697
Encompass
The Meso Scale Discovery R-PLEX Mouse MMP-3 Total Assay measures total MMP-3 in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity wide dynamic range and low sample volume The assay can be multiplexed with other R-PLEX assays The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls to quantify MMP-3 aiding in the analysis of its role in extracellular matrix remodeling
Catalog No. 50-000-78697 Supplier Meso Scale Discovery Supplier No. K152ZFR2
The Meso Scale Discovery R-PLEX Mouse MMP-3 Total Assay measures total MMP-3 in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity wide dynamic range and low sample volume The assay can be multiplexed with other R-PLEX assays The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls to quantify MMP-3 aiding in the analysis of its role in extracellular matrix remodeling