Learn More
Meso Scale Discovery R-PLEX Mouse Pentraxin 3 ASY
SDP
Catalog No. 5000079699
Encompass
The Meso Scale Discovery R-PLEX Mouse Pentraxin 3 Assay is a multiplex immunoassay that quantitatively measures mouse Pentraxin 3 in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The assay has a lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 ng/mL This assay allows for the rapid reproducible quantification of mouse Pentraxin 3 from various biological matrices
Catalog No. 50-000-79699 Supplier Meso Scale Discovery Supplier No. K1520MR2
The Meso Scale Discovery R-PLEX Mouse Pentraxin 3 Assay is a multiplex immunoassay that quantitatively measures mouse Pentraxin 3 in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The assay has a lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 ng/mL This assay allows for the rapid reproducible quantification of mouse Pentraxin 3 from various biological matrices