Learn More
Meso Scale Discovery R-PLEX Mouse Pentraxin 3 ASYQP
SDP
Catalog No. 5000079729
Encompass
The Meso Scale Discovery R-PLEX Mouse Pentraxin 3 Assay is a multiplex immunoassay kit for quantifying pentraxin 3 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision Requiring 50 L or less sample volume it can assay up to 80 samples in duplicates in 3 5 hours The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent enabling rapid reproducible quantification of pentraxin 3
Catalog No. 50-000-79729 Supplier Meso Scale Discovery Supplier No. K1520MR22
The Meso Scale Discovery R-PLEX Mouse Pentraxin 3 Assay is a multiplex immunoassay kit for quantifying pentraxin 3 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision Requiring 50 L or less sample volume it can assay up to 80 samples in duplicates in 3 5 hours The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent enabling rapid reproducible quantification of pentraxin 3