Learn More
Meso Scale Discovery R-PLEX Mouse PSP-D ASY
SDP
Catalog No. 5000081493
Encompass
The R-Plex Mouse PSP/D Assay from Meso Scale Discovery is a multiplex immunoassay measuring Prostate Specific Protein (PSP) and Prostatic Acid Phosphatase (PAP) in mouse serum and plasma Using electrochemiluminescence (ECL) detection it offers high sensitivity broad dynamic range and precision It allows simultaneous quantitative measurement of PSP and PAP biomarkers of prostate cancer in mouse models requiring low sample volume and providing results in 3 5 hours The assay is used to monitor cancer progression and evaluate therapeutic responses in preclinical studies
Catalog No. 50-000-81493 Supplier Meso Scale Discovery Supplier No. K1529XR2
The R-Plex Mouse PSP/D Assay from Meso Scale Discovery is a multiplex immunoassay measuring Prostate Specific Protein (PSP) and Prostatic Acid Phosphatase (PAP) in mouse serum and plasma Using electrochemiluminescence (ECL) detection it offers high sensitivity broad dynamic range and precision It allows simultaneous quantitative measurement of PSP and PAP biomarkers of prostate cancer in mouse models requiring low sample volume and providing results in 3 5 hours The assay is used to monitor cancer progression and evaluate therapeutic responses in preclinical studies