Learn More
Meso Scale Discovery R-PLEX Mouse PSP-D ASYQP
SDP
Catalog No. 5000078485
Encompass
The Meso Scale Discovery Mouse R-PLEX IL-2 Panel 22-Plex is a multiplex immunoassay that measures 22 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF CXCL1 CCL2 CCL3 CCL4 CCL5 CCL11 CXCL10 CXCL9 CXCL13 and VEGF) simultaneously from a single sample This assay allows high-throughput quantitative measurement of multiple analytes from a small sample volume using electrochemiluminescence detection on the MSD platform The panel is for research use only to better understand immune responses in mouse models
Catalog No. 50-000-78485 Supplier Meso Scale Discovery Supplier No. K1529XR22
The Meso Scale Discovery Mouse R-PLEX IL-2 Panel 22-Plex is a multiplex immunoassay that measures 22 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF CXCL1 CCL2 CCL3 CCL4 CCL5 CCL11 CXCL10 CXCL9 CXCL13 and VEGF) simultaneously from a single sample This assay allows high-throughput quantitative measurement of multiple analytes from a small sample volume using electrochemiluminescence detection on the MSD platform The panel is for research use only to better understand immune responses in mouse models