Learn More
Meso Scale Discovery R-PLEX Mouse/Rat TREM-1 ASYQP
SDP
Catalog No. 5000078365
Encompass
The Meso Scale Discovery Mouse R-Plex K150U1R-22 is a multiplex immunoassay panel for measuring 22 mouse biomarkers including IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF CXCL1 CCL2 CCL3 CCL4 CCL5 CCL11 CXCL10 VEGF Leptin and Resistin It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs Suitable for mouse serum plasma and cell culture supernatants the kit includes a 96-well plate with capture antibodies detection antibodies and calibrator solutions
Catalog No. 50-000-78365 Supplier Meso Scale Discovery Supplier No. K150U1R22
The Meso Scale Discovery Mouse R-Plex K150U1R-22 is a multiplex immunoassay panel for measuring 22 mouse biomarkers including IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF CXCL1 CCL2 CCL3 CCL4 CCL5 CCL11 CXCL10 VEGF Leptin and Resistin It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs Suitable for mouse serum plasma and cell culture supernatants the kit includes a 96-well plate with capture antibodies detection antibodies and calibrator solutions