Learn More
Meso Scale Discovery R-PLEX Mouse RBP4 ASY
SDP
Catalog No. 5000082043
Encompass
The R-PLEX Mouse RBP4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures RBP4 levels in mouse serum plasma and tissue culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents needed to perform the assay The kit can quantitatively measure RBP4 levels with high precision and accuracy It is suitable for studying RBP4 biology and evaluating RBP4 as a biomarker
Catalog No. 50-000-82043 Supplier Meso Scale Discovery Supplier No. K1522QR2
The R-PLEX Mouse RBP4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures RBP4 levels in mouse serum plasma and tissue culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluents needed to perform the assay The kit can quantitatively measure RBP4 levels with high precision and accuracy It is suitable for studying RBP4 biology and evaluating RBP4 as a biomarker