Learn More
Meso Scale Discovery R-PLEX Mouse RBP4 ASYQP
SDP
Catalog No. 5000079989
Encompass
The R-PLEX Mouse RBP4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures RBP4 levels in mouse serum plasma and tissue culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls The assay has a typical sensitivity of 1 pg/mL and an upper limit of quantification of 10 000 pg/mL for RBP4 It requires a small sample volume of 25 L and the total assay time is 3 5 hours
Catalog No. 50-000-79989 Supplier Meso Scale Discovery Supplier No. K1522QR22
The R-PLEX Mouse RBP4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures RBP4 levels in mouse serum plasma and tissue culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls The assay has a typical sensitivity of 1 pg/mL and an upper limit of quantification of 10 000 pg/mL for RBP4 It requires a small sample volume of 25 L and the total assay time is 3 5 hours