Learn More
Meso Scale Discovery R-PLEX Mouse Resistin ASYQP
SDP
Catalog No. 5000082127
Encompass
The Meso Scale Discovery R-PLEX Mouse Resistin Assay Kit is a multiplex immunoassay that quantitatively measures resistin in mouse serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82127 Supplier Meso Scale Discovery Supplier No. K152ZOR22
The Meso Scale Discovery R-PLEX Mouse Resistin Assay Kit is a multiplex immunoassay that quantitatively measures resistin in mouse serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL