Learn More
Meso Scale Discovery R-PLEX Mouse SAA ASY
SDP
Catalog No. 5000078454
Encompass
The R-PLEX Mouse SAA Assay Kit from Meso Scale Discovery is an immunoassay that quantitatively measures serum amyloid A (SAA) in mouse serum plasma and tissue culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and reagents required to perform the assay SAA is an acute phase protein and biomarker of inflammation This assay allows sensitive reproducible measurement of SAA levels in mouse samples
Catalog No. 50-000-78454 Supplier Meso Scale Discovery Supplier No. K1528KR2
The R-PLEX Mouse SAA Assay Kit from Meso Scale Discovery is an immunoassay that quantitatively measures serum amyloid A (SAA) in mouse serum plasma and tissue culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and reagents required to perform the assay SAA is an acute phase protein and biomarker of inflammation This assay allows sensitive reproducible measurement of SAA levels in mouse samples