Learn More
Meso Scale Discovery R-PLEX Mouse SAA ASYQP
SDP
Catalog No. 5000078455
Encompass
The R-PLEX Mouse SAA Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of serum amyloid A (SAA) in mouse serum plasma and tissue culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid sensitive and reproducible method for measuring SAA levels The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours allowing high-throughput analysis of samples This assay provides a useful tool for studying inflammatory conditions and responses in mouse models of human disease
Catalog No. 50-000-78455 Supplier Meso Scale Discovery Supplier No. K1528KR22
The R-PLEX Mouse SAA Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of serum amyloid A (SAA) in mouse serum plasma and tissue culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a rapid sensitive and reproducible method for measuring SAA levels The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours allowing high-throughput analysis of samples This assay provides a useful tool for studying inflammatory conditions and responses in mouse models of human disease