Learn More
Meso Scale Discovery R-PLEX Mouse Tau total ASYQP
SDP
Catalog No. 5000081376
Encompass
The Meso Scale Discovery R-Plex Mouse Tau Total Assay is an electrochemiluminescent immunoassay for quantifying total tau in mouse serum plasma and brain tissue lysates It uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid tau level quantification The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent The assay takes about 3 5 hours for 96 samples offering a sensitive high-throughput method for measuring tau in mouse biofluids and tissues
Catalog No. 50-000-81376 Supplier Meso Scale Discovery Supplier No. K1528ER22
The Meso Scale Discovery R-Plex Mouse Tau Total Assay is an electrochemiluminescent immunoassay for quantifying total tau in mouse serum plasma and brain tissue lysates It uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid tau level quantification The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent The assay takes about 3 5 hours for 96 samples offering a sensitive high-throughput method for measuring tau in mouse biofluids and tissues