Learn More
Meso Scale Discovery R-PLEX Mouse TSLP ASYQP
SDP
Catalog No. 5000078452
Encompass
The R-PLEX Mouse TSLP Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and a 10X read buffer The detection range is 1 15 to 1500 pg/mL for mouse TSLP
Catalog No. 50-000-78452 Supplier Meso Scale Discovery Supplier No. K152ALKR22
The R-PLEX Mouse TSLP Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and a 10X read buffer The detection range is 1 15 to 1500 pg/mL for mouse TSLP