Learn More
Meso Scale Discovery R-PLEX Mouse VCAM-1 ASYQP
SDP
Catalog No. 5000080019
Encompass
The R-PLEX Mouse VCAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures mouse VCAM-1 in serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This 5-plex assay kit includes a VCAM-1 standard and antibodies pre-coated on plates provided in a 96-well format The kit is for research use only
Catalog No. 50-000-80019 Supplier Meso Scale Discovery Supplier No. K1527PR22
The R-PLEX Mouse VCAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures mouse VCAM-1 in serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This 5-plex assay kit includes a VCAM-1 standard and antibodies pre-coated on plates provided in a 96-well format The kit is for research use only