Learn More
Meso Scale Discovery R-PLEX NHP CD177 ASYQP
SDP
Catalog No. 5000081095
Encompass
The Meso Scale Discovery Non-Human Primate CD177 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of CD177 in non-human primate samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81095 Supplier Meso Scale Discovery Supplier No. K156ALSR22
The Meso Scale Discovery Non-Human Primate CD177 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of CD177 in non-human primate samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay takes approximately 3 5 hours to complete