Learn More
Meso Scale Discovery R-PLEX Rat Albumin ASYQP
SDP
Catalog No. 5000082137
Encompass
The R-PLEX Rat Albumin Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of albumin in rat serum plasma urine and other biological samples The assay uses MSD GOLD technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and sample diluent The assay has a range of 1 56 - 400 ng/mL and takes approximately 3 5 hours to complete This product can aid research of kidney function endothelial permeability and other disease states in rat models
Catalog No. 50-000-82137 Supplier Meso Scale Discovery Supplier No. K1534VR22
The R-PLEX Rat Albumin Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of albumin in rat serum plasma urine and other biological samples The assay uses MSD GOLD technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and sample diluent The assay has a range of 1 56 - 400 ng/mL and takes approximately 3 5 hours to complete This product can aid research of kidney function endothelial permeability and other disease states in rat models