Learn More
Meso Scale Discovery R-PLEX Rat β-NGF Assay, 5 plate size, Singleplex format, 96-well plate, Plasma and Serum sample types, Dynamic range 0.2 - 6000 pg/mL, LLOD 0.2 pg/mL, For research use only
SDP
Catalog No. NC3497304
Encompass
R-PLEX Rat β-NGF Assay, 5 plate size, Singleplex format, 96-well plate, Plasma and Serum sample types, Dynamic range 0.2 - 6000 pg/mL, LLOD 0.2 pg/mL, For research use only
Catalog No. NC3497304 Supplier Meso Scale Discovery Supplier No. K153ANFR2
R-PLEX Rat β-NGF Assay, 5 plate size, Singleplex format, 96-well plate, Plasma and Serum sample types, Dynamic range 0.2 - 6000 pg/mL, LLOD 0.2 pg/mL, For research use only