Learn More
Meso Scale Discovery R-PLEX Rat EPO ASY
SDP
Catalog No. 5000082162
Encompass
The R-PLEX Rat EPO Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 0 17 pg/mL and the upper limit of quantitation is 1500 pg/mL
Catalog No. 50-000-82162 Supplier Meso Scale Discovery Supplier No. K1531GR2
The R-PLEX Rat EPO Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 0 17 pg/mL and the upper limit of quantitation is 1500 pg/mL