Learn More
Meso Scale Discovery R-PLEX Rat GM-CSF ASYQP
SDP
Catalog No. 5000078680
Encompass
The R-Plex Rat GM-CSF Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat granulocyte-macrophage colony-stimulating factor (GM-CSF) in serum plasma tissue culture media and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay can detect rat GM-CSF with a lower limit of detection of 1 52 pg/mL and an upper limit of quantitation of 1500 pg/mL The assay is provided in a 96-well plate format and includes all necessary reagents
Catalog No. 50-000-78680 Supplier Meso Scale Discovery Supplier No. K1531CR22
The R-Plex Rat GM-CSF Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat granulocyte-macrophage colony-stimulating factor (GM-CSF) in serum plasma tissue culture media and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay can detect rat GM-CSF with a lower limit of detection of 1 52 pg/mL and an upper limit of quantitation of 1500 pg/mL The assay is provided in a 96-well plate format and includes all necessary reagents