Learn More
Meso Scale Discovery R-PLEX Rat HAVCR1/KIM-1 ASYQP
SDP
Catalog No. 5000082046
Encompass
The R-PLEX Rat HAVCR1 (KIM-1) Assay from Meso Scale Diagnostics is a 96-well electrochemiluminescent immunoassay for measuring Kidney Injury Molecule-1 (KIM-1) in rat serum plasma urine and tissue culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid KIM-1 quantification a key biomarker of kidney injury The kit includes a 96-well plate with anti-KIM-1 capture antibody detection antibody read buffer sample diluent calibrators and controls
Catalog No. 50-000-82046 Supplier Meso Scale Discovery Supplier No. K1534UR22
The R-PLEX Rat HAVCR1 (KIM-1) Assay from Meso Scale Diagnostics is a 96-well electrochemiluminescent immunoassay for measuring Kidney Injury Molecule-1 (KIM-1) in rat serum plasma urine and tissue culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid KIM-1 quantification a key biomarker of kidney injury The kit includes a 96-well plate with anti-KIM-1 capture antibody detection antibody read buffer sample diluent calibrators and controls