Learn More
Meso Scale Discovery R-PLEX Rat IFN- ASYQP
SDP
Catalog No. 5000080418
Encompass
The R-PLEX Rat IFN-gamma Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring rat interferon gamma (IFN-) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibody ECL-labeled detection antibody assay diluent wash buffer read buffer and IFN- standards It detects rat IFN- from 0 71 pg/mL to 2500 pg/mL
Catalog No. 50-000-80418 Supplier Meso Scale Discovery Supplier No. K1531RR22
The R-PLEX Rat IFN-gamma Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring rat interferon gamma (IFN-) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibody ECL-labeled detection antibody assay diluent wash buffer read buffer and IFN- standards It detects rat IFN- from 0 71 pg/mL to 2500 pg/mL